siRNA Details
| Gene Symbol: Snrpc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271040.1 | NP_001257969.1 | 2, 3 | 390 | ||
| XM_006256220.2 | XP_006256282.1 | 3, 4 | 449 | |||
| Gene Symbol: LOC685273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001063116.5 | XP_001063116.2 | 1 | 67 | ||

Snrpc
Genomic Map