siRNA Details
| Gene Symbol: Ddo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109465.2 | NP_001102935.1 | 4, 5 | 518 | ||
| GenBank mRNA | AB983592.1 | - | 3, 4 | 458 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddo| Gene Symbol: Ddo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109465.2 | NP_001102935.1 | 4, 5 | 518 | ||
| GenBank mRNA | AB983592.1 | - | 3, 4 | 458 | ||