siRNA Details
| Gene Symbol: Rpp30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191083.1 | NP_001178012.1 | 3 | 192 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rpp30| Gene Symbol: Rpp30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191083.1 | NP_001178012.1 | 3 | 192 | ||