siRNA Details
| Gene Symbol: LOC102548397 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763971.1 | XP_008762193.1 | 1 | 828 | ||
| Gene Symbol: Ankrd13c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191570.1 | NP_001178499.1 | 5 | 871 | ||
| XM_006233547.2 | XP_006233609.1 | 4 | 1796 | |||
| XM_008761571.1 | XP_008759793.1 | 3 | 1692 | |||

LOC102548397
Genomic Map