siRNA Details
| Gene Symbol: Arl14epl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006254700.2 | XP_006254762.1 | 3 | 325 | ||
| XM_006254701.2 | XP_006254763.1 | 3 | 409 | |||
| XM_006254702.2 | XP_006254764.1 | 2 | 216 | |||
| XM_008772183.1 | XP_008770405.1 | 3 | 488 | |||
| XM_008772184.1 | XP_008770406.1 | 3 | 270 | |||
| XM_008772185.1 | XP_008770407.1 | 2 | 224 | |||

Arl14epl
Genomic Map