siRNA Details
| Gene Symbol: Arl14epl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006254700.2 | XP_006254762.1 | 4 | 455 | ||
| XM_006254701.2 | XP_006254763.1 | 4 | 539 | |||
| XM_006254702.2 | XP_006254764.1 | 3 | 346 | |||
| XM_008772183.1 | XP_008770405.1 | 4 | 618 | |||
| XM_008772184.1 | XP_008770406.1 | 4 | 400 | |||
| XM_008772185.1 | XP_008770407.1 | 3 | 354 | |||

Arl14epl
Genomic Map