siRNA Details
Gene Symbol: Arl14epl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006254700.2 | XP_006254762.1 | 4 | 398 | ||
XM_006254701.2 | XP_006254763.1 | 4 | 482 | |||
XM_006254702.2 | XP_006254764.1 | 3 | 289 | |||
XM_008772183.1 | XP_008770405.1 | 4 | 561 | |||
XM_008772184.1 | XP_008770406.1 | 4 | 343 | |||
XM_008772185.1 | XP_008770407.1 | 3 | 297 |
