siRNA Details
Gene Symbol: Ppp1r26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001063635.4 | XP_001063635.1 | 4 | 2239 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ppp1r26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001063635.4 | XP_001063635.1 | 4 | 2239 |