siRNA Details
| Gene Symbol: LOC685406 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047977.1 | NP_001041442.1 | 4 | 289 | ||
| GenBank mRNA | AY387048.1 | - | 4 | 289 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC685406| Gene Symbol: LOC685406 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047977.1 | NP_001041442.1 | 4 | 289 | ||
| GenBank mRNA | AY387048.1 | - | 4 | 289 | ||