siRNA Details
| Gene Symbol: Evi2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044287.1 | NP_001037752.1 | 2 | 207 | ||
| GenBank mRNA | BC088856.1 | - | 2 | 207 | 7318412 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Evi2a| Gene Symbol: Evi2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044287.1 | NP_001037752.1 | 2 | 207 | ||
| GenBank mRNA | BC088856.1 | - | 2 | 207 | 7318412 | |