siRNA Details
| Gene Symbol: Emid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109467.1 | NP_001102937.1 | 8 | 940 | ||
| XM_008770367.1 | XP_008768589.1 | 15 | 1317 | |||
| XM_008770368.1 | XP_008768590.1 | 15 | 1311 | |||
| XM_008770369.1 | XP_008768591.1 | 16 | 1372 | |||
| XM_008770371.1 | XP_008768593.1 | 14 | 1244 | |||

Emid1
Genomic Map