siRNA Details
| Gene Symbol: Zfp385a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109470.2 | NP_001102940.1 | 4 | 313 | ||
| NM_001135088.1 | NP_001128560.1 | 4 | 465 | |||
| NM_001135089.1 | NP_001128561.1 | 4 | 364 | |||
| XM_006242445.2 | XP_006242507.1 | 3 | 490 | |||
| XM_006242446.2 | XP_006242508.1 | 4 | 670 | |||
| GenBank mRNA | BC166729.1 | - | 4 | 465 | 7934026 | |

Zfp385a
Genomic Map