siRNA Details
| Gene Symbol: Rbbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107912.2 | NP_001101382.1 | 12 | 1723 | ||
| GenBank mRNA | BC168994.1 | - | 12 | 1723 | 8362559 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbbp4| Gene Symbol: Rbbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107912.2 | NP_001101382.1 | 12 | 1723 | ||
| GenBank mRNA | BC168994.1 | - | 12 | 1723 | 8362559 | |