siRNA Details
| Gene Symbol: Potea | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770390.1 | XP_008768612.1 | 26 | 4080 | ||
| XM_008770391.1 | XP_008768613.1 | 26 | 4059 | |||
| XM_008770392.1 | XP_008768614.1 | 25 | 3906 | |||
| XM_008770393.1 | XP_008768615.1 | 21 | 4822 | |||

Potea
Genomic Map