siRNA Details
| Gene Symbol: Snrpe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271237.1 | NP_001258166.1 | 3 | 149 | ||
| GenBank mRNA | FQ211122.1 | - | 3 | 149 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snrpe| Gene Symbol: Snrpe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271237.1 | NP_001258166.1 | 3 | 149 | ||
| GenBank mRNA | FQ211122.1 | - | 3 | 149 | ||