siRNA Details
Gene Symbol: Pik3ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133399.2 | NP_596890.2 | 19 | 2880 | ||
GenBank mRNA | AF395897.1 | - | 7 | 768 |
Gene Symbol: LOC685590 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773148.1 | XP_008771370.1 | 3 | 3000 |
