siRNA Details
| Gene Symbol: MLEC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303627.1 | NP_001290556.1 | 3 | 357 | ||
| NM_014730.3 | NP_055545.1 | 3 | 640 | |||
| XM_011539031.1 | XP_011537333.1 | 3 | 5055 | |||
| XM_011539032.1 | XP_011537334.1 | 4 | 4743 | |||
| GenBank mRNA | BC000371.2 | - | 3 | 589 | 2819615 | |
| BC016297.2 | - | 3 | 591 | 3832682 | ||
| BI458950.1 | - | 3 | 359 | |||
| BU845446.1 | - | 618 | ||||
| D63486.1 | - | 3 | 595 | |||

MLEC
Genomic Map