siRNA Details
| Gene Symbol: Zfp59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759213.1 | XP_008757435.1 | 5 | 1906 | ||
| XM_008759214.1 | XP_008757436.1 | 5 | 1423 | |||
| XM_008759215.1 | XP_008757437.1 | 2 | 151 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp59| Gene Symbol: Zfp59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759213.1 | XP_008757435.1 | 5 | 1906 | ||
| XM_008759214.1 | XP_008757436.1 | 5 | 1423 | |||
| XM_008759215.1 | XP_008757437.1 | 2 | 151 | |||