siRNA Details
| Gene Symbol: LOC685590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773148.1 | XP_008771370.1 | 3 | 1237 | ||
| Gene Symbol: Pik3ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133399.2 | NP_596890.2 | 5 | 1111 | ||
| GenBank mRNA | CK366305.1 | - | 3 | 343 | ||

LOC685590
Genomic Map