siRNA Details
Gene Symbol: Pik3ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133399.2 | NP_596890.2 | 5 | 1130 | ||
GenBank mRNA | CK366305.1 | - | 3 | 362 |
Gene Symbol: LOC685590 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773148.1 | XP_008771370.1 | 3 | 1256 |
