siRNA Details
| Gene Symbol: KIAA0513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286565.1 | NP_001273494.1 | 3 | 1037 | ||
| NM_001286566.1 | NP_001273495.1 | 3 | 686 | |||
| NM_001297766.1 | NP_001284695.1 | 3 | 679 | |||
| NM_014732.3 | NP_055547.1 | 3 | 679 | |||
| XM_005256265.3 | XP_005256322.1 | 3 | 728 | |||
| XM_017023912.1 | XP_016879401.1 | 3 | 3254 | |||
| GenBank mRNA | AB011085.1 | - | 3 | 1037 | ||
| AK299892.1 | - | 3 | 670 | |||
| BC030280.1 | - | 3 | 626 | 5211581 | ||
| BU844795.1 | - | 3 | 698 | |||

KIAA0513
Genomic Map