siRNA Details
| Gene Symbol: Dnal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109477.2 | NP_001102947.2 | 5 | 344 | ||
| XM_006240352.2 | XP_006240414.1 | 6 | 553 | |||
| GenBank mRNA | BC161941.1 | - | 383 | |||
| EV765393.1 | - | 6 | 383 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnal1| Gene Symbol: Dnal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109477.2 | NP_001102947.2 | 5 | 344 | ||
| XM_006240352.2 | XP_006240414.1 | 6 | 553 | |||
| GenBank mRNA | BC161941.1 | - | 383 | |||
| EV765393.1 | - | 6 | 383 | |||