siRNA Details
| Gene Symbol: Fam122c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006257649.2 | XP_006257711.1 | 2 | 207 | ||
| XM_006257651.2 | XP_006257713.1 | 2 | 207 | |||
| XM_008773640.1 | XP_008771862.1 | 2 | 207 | |||
| XM_008773641.1 | XP_008771863.1 | 2 | 243 | |||
| XM_008773642.1 | XP_008771864.1 | 2 | 207 | |||
| XM_008773643.1 | XP_008771865.1 | 2 | 207 | |||
| XM_008773644.1 | XP_008771866.1 | 2 | 207 | |||

Fam122c
Genomic Map