siRNA Details
| Gene Symbol: Ggct | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108629.2 | NP_001102099.1 | 2 | 342 | ||
| XM_008762899.1 | XP_008761121.1 | 2 | 396 | |||
| GenBank mRNA | FM060344.1 | - | 2 | 280 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ggct| Gene Symbol: Ggct | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108629.2 | NP_001102099.1 | 2 | 342 | ||
| XM_008762899.1 | XP_008761121.1 | 2 | 396 | |||
| GenBank mRNA | FM060344.1 | - | 2 | 280 | ||