siRNA Details
| Gene Symbol: Rpl26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006246606.2 | XP_006246668.1 | 2 | 267 | ||
| GenBank mRNA | FQ209942.1 | - | 2 | 113 | ||
| FQ210265.1 | - | 2 | 113 | |||
| FQ210305.1 | - | 2 | 114 | |||
| FQ211391.1 | - | 2 | 136 | |||
| FQ214577.1 | - | 2 | 113 | |||
| FQ218083.1 | - | 2 | 113 | |||
| FQ218115.1 | - | 2 | 112 | |||
| FQ220102.1 | - | 113 | ||||
| FQ221223.1 | - | 2 | 115 | |||
| FQ221302.1 | - | 2 | 113 | |||
| FQ221303.1 | - | 2 | 113 | |||
| FQ221304.1 | - | 2 | 113 | |||
| FQ221369.1 | - | 2 | 113 | |||
| FQ221596.1 | - | 2 | 108 | |||
| FQ221712.1 | - | 2 | 113 | |||
| FQ221762.1 | - | 2 | 113 | |||
| FQ222089.1 | - | 2 | 113 | |||
| FQ222135.1 | - | 2 | 113 | |||
| FQ222176.1 | - | 2 | 113 | |||
| FQ222323.1 | - | 2 | 107 | |||
| FQ222676.1 | - | 2 | 113 | |||
| FQ222872.1 | - | 2 | 113 | |||
| FQ222877.1 | - | 2 | 107 | |||
| FQ222953.1 | - | 2 | 113 | |||
| FQ223076.1 | - | 2 | 114 | |||
| FQ223161.1 | - | 2 | 113 | |||
| FQ223442.1 | - | 2 | 113 | |||
| FQ224611.1 | - | 2 | 113 | |||
| FQ228453.1 | - | 2 | 112 | |||
| FQ228549.1 | - | 2 | 107 | |||
| FQ228592.1 | - | 2 | 107 | |||
| FQ228636.1 | - | 2 | 113 | |||
| FQ228935.1 | - | 2 | 114 | |||
| X14671.1 | - | 2 | 108 | |||

Rpl26
Genomic Map