siRNA Details
| Gene Symbol: Hoxa6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191087.1 | NP_001178016.1 | 265 | |||
| Gene Symbol: LOC100909604 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762945.1 | XP_008761167.1 | 1 | 4696 | ||
| Gene Symbol: LOC100911511 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003752179.3 | XP_003752227.1 | 1 | 2641 | ||

Hoxa6
Genomic Map