siRNA Details
| Gene Symbol: LOC100909604 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762944.1 | XP_008761166.1 | 2 | 2708 | ||
| XM_008762945.1 | XP_008761167.1 | 1 | 4957 | |||
| Gene Symbol: Hoxa6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191087.1 | NP_001178016.1 | 526 | |||
| XM_006236513.2 | XP_006236575.1 | 2 | 2085 | |||
| Gene Symbol: LOC100911511 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003752179.3 | XP_003752227.1 | 1 | 2902 | ||
| XM_006257703.2 | XP_006257765.1 | 2 | 2708 | |||

LOC100909604
Genomic Map