siRNA Details
| Gene Symbol: Phemx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728846.4 | XP_002728892.3 | 6 | 2881 | ||
| XM_006230915.2 | XP_006230977.1 | 5 | 2806 | |||
| GenBank mRNA | BC158837.1 | - | 4 | 346 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phemx| Gene Symbol: Phemx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728846.4 | XP_002728892.3 | 6 | 2881 | ||
| XM_006230915.2 | XP_006230977.1 | 5 | 2806 | |||
| GenBank mRNA | BC158837.1 | - | 4 | 346 | ||