siRNA Details
| Gene Symbol: LOC685767 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001065185.5 | XP_001065185.4 | 3 | 607 | ||
| XM_008768748.1 | XP_008766970.1 | 3 | 756 | |||
| XM_008768749.1 | XP_008766971.1 | 3 | 756 | |||
| XM_008768750.1 | XP_008766972.1 | 3 | 756 | |||
| XM_008768751.1 | XP_008766973.1 | 3 | 756 | |||
| XM_008768752.1 | XP_008766974.1 | 3 | 756 | |||
| XM_008768753.1 | XP_008766975.1 | 2 | 417 | |||

LOC685767
Genomic Map