siRNA Details
| Gene Symbol: KIAA0101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029989.2 | NP_001025160.1 | 2 | 244 | ||
| NM_014736.5 | NP_055551.1 | 2 | 244 | |||
| NR_109934.1 | 2 | 300 | ||||
| GenBank mRNA | AF529370.1 | - | 2 | 105 | ||
| AK290748.1 | - | 2 | 175 | |||
| AV757440.1 | - | 2 | 274 | |||
| AY598324.1 | - | 2 | 105 | |||
| BC005832.2 | - | 2 | 164 | 2988995 | ||
| BC007101.1 | - | 2 | 171 | 4281110 | ||
| BC016782.1 | - | 2 | 179 | 4109322 | ||
| BF691807.1 | - | 2 | 245 | |||
| BG502841.1 | - | 237 | ||||
| D14657.1 | - | 2 | 166 | |||
| DT220867.1 | - | 2 | 297 | |||

KIAA0101
Genomic Map