siRNA Details
| Gene Symbol: LOC685767 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001065185.5 | XP_001065185.4 | 4 | 993 | ||
| XM_008768748.1 | XP_008766970.1 | 4 | 1142 | |||
| XM_008768753.1 | XP_008766975.1 | 3 | 803 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC685767| Gene Symbol: LOC685767 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001065185.5 | XP_001065185.4 | 4 | 993 | ||
| XM_008768748.1 | XP_008766970.1 | 4 | 1142 | |||
| XM_008768753.1 | XP_008766975.1 | 3 | 803 | |||