siRNA Details
| Gene Symbol: Ddr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031764.3 | NP_113952.2 | 11 | 1584 | ||
| XM_006250226.2 | XP_006250288.1 | 12 | 4881 | |||
| XM_006250227.2 | XP_006250289.1 | 14 | 2216 | |||
| XM_006250228.2 | XP_006250290.1 | 13 | 1997 | |||
| XM_006250229.2 | XP_006250291.1 | 13 | 1945 | |||
| XM_008769761.1 | XP_008767983.1 | 13 | 1959 | |||
| XM_008769762.1 | XP_008767984.1 | 13 | 1891 | |||
| XM_008769763.1 | XP_008767985.1 | 13 | 2197 | |||
| GenBank mRNA | BC161961.1 | - | 11 | 1584 | 9027893 | |

Ddr2
Genomic Map