siRNA Details
| Gene Symbol: Cnpy2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077585.1 | NP_001071053.1 | 5 | 755 | ||
| GenBank mRNA | BC126101.1 | - | 5 | 755 | 7319422 | |
| FQ220100.1 | - | 5 | 611 | |||
| FQ220178.1 | - | 5 | 739 | |||
| FQ225671.1 | - | 5 | 691 | |||
| FQ231871.1 | - | 5 | 662 | |||

Cnpy2
Genomic Map