siRNA Details
| Gene Symbol: Enthd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135913.1 | NP_001129385.1 | 7 | 1760 | ||
| XM_006242122.2 | XP_006242184.1 | 8 | 2257 | |||
| XM_008765787.1 | XP_008764009.1 | 8 | 2290 | |||
| XM_008765788.1 | XP_008764010.1 | 8 | 2257 | |||
| XM_008765789.1 | XP_008764011.1 | 6 | 1476 | |||
| XM_008765790.1 | XP_008764012.1 | 6 | 2155 | |||
| XM_008765791.1 | XP_008764013.1 | 6 | 1440 | |||
| GenBank mRNA | BC167764.1 | - | 7 | 1760 | 7453894 | |

Enthd1
Genomic Map