siRNA Details
| Gene Symbol: Rprml | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109481.1 | NP_001102951.1 | 1 | 923 | ||
| GenBank mRNA | FQ220063.1 | - | 1 | 953 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rprml| Gene Symbol: Rprml | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109481.1 | NP_001102951.1 | 1 | 923 | ||
| GenBank mRNA | FQ220063.1 | - | 1 | 953 | ||