siRNA Details
| Gene Symbol: Dnajb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130510.1 | NP_001123982.1 | 1 | 547 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnajb7| Gene Symbol: Dnajb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130510.1 | NP_001123982.1 | 1 | 547 | ||