siRNA Details
Gene Symbol: Jazf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001065610.5 | XP_001065610.1 | 3 | 388 | ||
XM_006236535.2 | XP_006236597.1 | 3 | 1176 | |||
XM_006236536.2 | XP_006236598.1 | 3 | 2294 | |||
XM_008762953.1 | XP_008761175.1 | 3 | 515 | |||
XM_008762954.1 | XP_008761176.1 | 4 | 730 |
