siRNA Details
| Gene Symbol: Hnrnpa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY363226.1 | - | 4 | 398 | ||
| AY363227.1 | - | 4 | 332 | |||
| CB725005.1 | - | 4 | 324 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hnrnpa3| Gene Symbol: Hnrnpa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY363226.1 | - | 4 | 398 | ||
| AY363227.1 | - | 4 | 332 | |||
| CB725005.1 | - | 4 | 324 | |||