siRNA Details
| Gene Symbol: Uroc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001066120.5 | XP_001066120.2 | 8 | 827 | ||
| XM_008763138.1 | XP_008761360.1 | 8 | 1254 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uroc1| Gene Symbol: Uroc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001066120.5 | XP_001066120.2 | 8 | 827 | ||
| XM_008763138.1 | XP_008761360.1 | 8 | 1254 | |||