siRNA Details
| Gene Symbol: Tnfrsf22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006230917.1 | XP_006230979.1 | 5 | 703 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnfrsf22| Gene Symbol: Tnfrsf22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006230917.1 | XP_006230979.1 | 5 | 703 | ||