siRNA Details
| Gene Symbol: Casq1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159594.1 | NP_001153066.1 | 1 | 315 | ||
| GenBank mRNA | CA334109.1 | - | 1 | 315 | ||
| FQ224875.1 | - | 2 | 142 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Casq1| Gene Symbol: Casq1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159594.1 | NP_001153066.1 | 1 | 315 | ||
| GenBank mRNA | CA334109.1 | - | 1 | 315 | ||
| FQ224875.1 | - | 2 | 142 | |||