siRNA Details
| Gene Symbol: Btnl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166351.1 | NP_001159823.1 | 5 | 928 | ||
| Gene Symbol: LOC100912021 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772784.1 | XP_008771006.1 | 4 | 811 | ||

Btnl5
Genomic Map