siRNA Details
| Gene Symbol: KIAA0232 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100590.1 | NP_001094060.1 | 6 | 1881 | ||
| NM_014743.2 | NP_055558.2 | 7 | 1965 | |||
| XM_005248041.4 | XP_005248098.1 | 4 | 1440 | |||
| XM_011513598.2 | XP_011511900.1 | 7 | 1618 | |||
| XM_017008860.1 | XP_016864349.1 | 6 | 2911 | |||
| XM_017008861.1 | XP_016864350.1 | 5 | 1628 | |||
| XM_017008862.1 | XP_016864351.1 | 6 | 2266 | |||
| XM_017008863.1 | XP_016864352.1 | 5 | 1628 | |||
| GenBank mRNA | AK124715.1 | - | 6 | 1821 | ||
| BC150286.1 | - | 7 | 1957 | 8860111 | ||
| D86985.3 | - | 7 | 1957 | |||
| EF553521.1 | - | 6 | 1822 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001081232, BC094673, AK122224, AK152987, AK134363, and others.

KIAA0232
Genomic Map