siRNA Details
| Gene Symbol: Nsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107337.1 | NP_001100807.1 | 23 | 7799 | ||
| XM_006253620.2 | XP_006253682.1 | 23 | 8048 | |||
| XM_006253621.2 | XP_006253683.1 | 23 | 8150 | |||
| XM_006253622.2 | XP_006253684.1 | 23 | 7878 | |||
| XM_006253623.2 | XP_006253685.1 | 23 | 8037 | |||
| XM_006253624.2 | XP_006253686.1 | 23 | 7974 | |||
| XM_006253625.2 | XP_006253687.1 | 23 | 8153 | |||
| XM_008771491.1 | XP_008769713.1 | 23 | 8103 | |||
| XM_008771492.1 | XP_008769714.1 | 22 | 7014 | |||
| XM_008771493.1 | XP_008769715.1 | 22 | 7304 | |||

Nsd1
Genomic Map