siRNA Details
| Gene Symbol: Gapt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109491.1 | NP_001102961.1 | 2 | 693 | ||
| XM_006231916.2 | XP_006231978.1 | 2 | 769 | |||
| XM_006231917.2 | XP_006231979.1 | 2 | 729 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gapt| Gene Symbol: Gapt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109491.1 | NP_001102961.1 | 2 | 693 | ||
| XM_006231916.2 | XP_006231978.1 | 2 | 769 | |||
| XM_006231917.2 | XP_006231979.1 | 2 | 729 | |||