siRNA Details
| Gene Symbol: Ccdc144b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760976.1 | XP_008759198.1 | 6 | 743 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc144b| Gene Symbol: Ccdc144b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760976.1 | XP_008759198.1 | 6 | 743 | ||