siRNA Details
Gene Symbol: Fam47e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770095.1 | XP_008768317.1 | 8 | 1268 | ||
XM_008770096.1 | XP_008768318.1 | 7 | 1175 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam47e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770095.1 | XP_008768317.1 | 8 | 1268 | ||
XM_008770096.1 | XP_008768318.1 | 7 | 1175 |