siRNA Details
| Gene Symbol: LOC686142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751717.3 | XP_003751765.1 | 5 | 696 | ||
| XM_006254133.2 | XP_006254195.1 | 4 | 1299 | |||
| XM_006254135.2 | XP_006254197.1 | 4 | 440 | |||
| Gene Symbol: LOC102553208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006254077.2 | XP_006254139.1 | 4 | 658 | ||
| XM_006254078.2 | XP_006254140.1 | 4 | 811 | |||

LOC686142
Genomic Map