siRNA Details
| Gene Symbol: Styx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271541.1 | NP_001258470.1 | 9 | 653 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Styx| Gene Symbol: Styx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271541.1 | NP_001258470.1 | 9 | 653 | ||