siRNA Details
Gene Symbol: Ttc39c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077231.1 | NP_001070699.1 | 3 | 235 | ||
XM_006254423.2 | XP_006254485.1 | 3 | 993 | |||
XM_008771954.1 | XP_008770176.1 | 2 | 119 | |||
GenBank mRNA | BC105623.1 | - | 3 | 235 | 7455888 |
